3-Bromo-6-fluorobenzene -1,2-diamine 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Bromo-2,3-diamino-4-fluorobenzene
PC501049-1G
PC501049-250MG
PC501049-5G
APOSPC501049-1GEA
14190
CZK
APOSPC501049-1G
APOSPC501049-250MG
APOSPC501049-5G
3-Bromo-6-fluorobenzene -1,2-diamine 95%
3-Bromo-6-fluorobenzene -1,2-diamine
Vzorec:
C₆H₆BrFN₂ Storage Temperature: Ambient |
MDL Number:
MFCD28154969 CAS číslo: 1805502-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...