4-Bromo-5-fluoro-2-(trifluoromethyl)-1H-benzimidazole
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC501009-1G
PC501009-250MG
APOSPC501009-1GEA
17360
CZK
APOSPC501009-1G
APOSPC501009-250MG
4-Bromo-5-fluoro-2-(trifluoromethyl)-1H-benzimidazole
4-Bromo-5-fluoro-2-(trifluoromethyl)-1H-benzimidazole
Vzorec:
C₈H₃BrF₄N₂ MW: 283,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28154938 CAS číslo: 1822861-98-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...