4-Chloro-6-fluoro-1H-benzimidazole
Dodavatel: Apollo Scientific
PC500985-1G
PC500985-5G
APOSPC500985-1GEA
4730
CZK
APOSPC500985-1G
APOSPC500985-5G
4-Chloro-6-fluoro-1H-benzimidazole
4-Chloro-6-fluoro-1H-benzimidazole
Vzorec:
C₇H₄ClFN₂ MW: 170,571 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22493415 CAS číslo: 1314092-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...