Tisk…

3-(2,2-Difluoroethoxy)benzyl alcohol 95%

Dodavatel: Apollo Scientific
Varování

PC500934-1G PC500934-250MG PC500934-5G
APOSPC500934-1GEA 9100 CZK
APOSPC500934-1G APOSPC500934-250MG APOSPC500934-5G
3-(2,2-Difluoroethoxy)benzyl alcohol 95%
3-(2,2-Difluoroethoxy)benzyl alcohol
Vzorec: C₉H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12807610
CAS číslo: 1184667-43-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR