3-(2,2-Difluoroethoxy)benzaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC500930-1G
PC500930-250MG
PC500930-5G
APOSPC500930-1GEA
10980
CZK
APOSPC500930-1G
APOSPC500930-250MG
APOSPC500930-5G
3-(2,2-Difluoroethoxy)benzaldehyde 95%
3-(2,2-Difluoroethoxy)benzaldehyde
Vzorec:
C₉H₈F₂O₂ MW: 186,155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12807548 CAS číslo: 467458-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...