Tisk…

3-(2,2,2-Trifluoroethoxy)benzoic acid

Dodavatel: Apollo Scientific

PC500842-1G PC500842-5G
APOSPC500842-1GEA 8200 CZK
APOSPC500842-1G APOSPC500842-5G
3-(2,2,2-Trifluoroethoxy)benzoic acid
3-(2,2,2-Trifluoroethoxy)benzoic acid
Vzorec: C₉H₇F₃O₃
MW: 220,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04967196
CAS číslo: 35480-48-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR