Tisk…

Methyl 2-chloro-5-(trifluoromethyl)isonicotinate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-4-(methoxycarbonyl)-5-(trifluoromethyl)pyridine, Methyl 2-chloro-5-(trifluoromethyl)pyridine-4-carboxylate

PC50076-1G PC50076-250MG
APOSPC50076-1GEA 6880 CZK
APOSPC50076-1G APOSPC50076-250MG
Methyl 2-chloro-5-(trifluoromethyl)isonicotinate 98%
Methyl 2-chloro-5-(trifluoromethyl)isonicotinate
Vzorec: C₈H₅ClF₃NO₂
MW: 239,579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18258738
CAS číslo: 1246685-28-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR