Tisk…

4-(Bromomethyl)-3-fluorobenzeneboronic acid 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC500745-1G PC500745-250MG PC500745-5G
APOSPC500745-1GEA 9920 CZK
APOSPC500745-1G APOSPC500745-250MG APOSPC500745-5G
4-(Bromomethyl)-3-fluorobenzeneboronic acid 95%
4-(Bromomethyl)-3-fluorobenzeneboronic acid
Vzorec: C₇H₇BBrFO₂
MW: 232,843 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28122909
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR