3-(Bromomethyl)-2-fluorobenzeneboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC500736-1G
PC500736-5G
APOSPC500736-1GEA
7440
CZK
APOSPC500736-1G
APOSPC500736-5G
3-(Bromomethyl)-2-fluorobenzeneboronic acid
3-(Bromomethyl)-2-fluorobenzeneboronic acid
Vzorec:
C₇H₇BBrFO₂ MW: 232,843 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28122904 CAS číslo: 680610-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...