3-Bromo-6-chloro-2-fluorobenzeneboronic acid
Dodavatel: Apollo Scientific
Synonyms:
3-Bromo-6-Chloro-2-fluorophenylboronic acid
PC500712-1G
PC500712-5G
APOSPC500712-1GEA
5590
CZK
APOSPC500712-1G
APOSPC500712-5G
3-Bromo-6-chloro-2-fluorobenzeneboronic acid
3-Bromo-6-chloro-2-fluorobenzeneboronic acid
Vzorec:
C₆H₄BBrClFO₂ MW: 253,261 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11044930 CAS číslo: 1451393-00-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...