Tisk…

4-Iodo-3-(trifluoromethyl)benzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC500683-1G PC500683-250MG PC500683-5G
APOSPC500683-1GEA 1660 CZK
APOSPC500683-1G APOSPC500683-250MG APOSPC500683-5G
4-Iodo-3-(trifluoromethyl)benzyl bromide 95%
4-Iodo-3-(trifluoromethyl)benzyl bromide
Vzorec: C₈H₅BrF₃I
MW: 364.929 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398194
CAS číslo: 1261457-09-3
UN: 3261
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR