4-Iodo-3-(trifluoromethyl)benzyl alcohol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC500677-1G
PC500677-250MG
PC500677-5G
APOSPC500677-1GEA
1660
CZK
APOSPC500677-1G
APOSPC500677-250MG
APOSPC500677-5G
4-Iodo-3-(trifluoromethyl)benzyl alcohol 95%
4-Iodo-3-(trifluoromethyl)benzyl alcohol
Vzorec:
C₈H₆F₃IO MW: 302,032 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18398177 CAS číslo: 959632-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...