2-(Bromomethyl)-5-fluorobenzenesulphonyl chloride 90%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-(Chlorosulphonyl)-4-fluorobenzyl bromide
PC500582-1G
PC500582-250MG
APOSPC500582-1GEA
11380
CZK
APOSPC500582-1G
APOSPC500582-250MG
2-(Bromomethyl)-5-fluorobenzenesulphonyl chloride 90%
2-(Bromomethyl)-5-fluorobenzenesulphonyl chloride
Vzorec:
C₇H₅BrClFO₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD28096593 CAS číslo: 1820717-41-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...