Tisk…

5-Bromo-2-fluoro-4-methylphenol

Dodavatel: Apollo Scientific
Varování

PC500577-1G PC500577-5G
APOSPC500577-1GEA 3480 CZK
APOSPC500577-1G APOSPC500577-5G
5-Bromo-2-fluoro-4-methylphenol
5-Bromo-2-fluoro-4-methylphenol
Vzorec: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD12405415
CAS číslo: 1111096-04-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR