Tisk…

3-(Bromomethyl)-2-fluorobenzaldehyde

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Fluoro-3-formylbenzyl bromide

PC500570-1G PC500570-250MG
APOSPC500570-1GEA 15480 CZK
APOSPC500570-1G APOSPC500570-250MG
3-(Bromomethyl)-2-fluorobenzaldehyde
3-(Bromomethyl)-2-fluorobenzaldehyde
Vzorec: C₈H₆BrFO
MW: 217,035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18378584
CAS číslo: 1379307-88-6
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR