3-(Bromomethyl)-2-fluorobenzonitrile
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC500556-1G
PC500556-250MG
PC500556-5G
APOSPC500556-1GEA
6550
CZK
APOSPC500556-1G
APOSPC500556-250MG
APOSPC500556-5G
3-(Bromomethyl)-2-fluorobenzonitrile
3-(Bromomethyl)-2-fluorobenzonitrile
Vzorec:
C₈H₅BrFN Storage Temperature: Refrigerator |
MDL Number:
MFCD18394043 CAS číslo: 1246527-18-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...