Tisk…

2-(Bromomethyl)-4-fluorobenzaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Fluoro-2-formylbenzyl bromide, α-Bromo-4-fluoro-o-tolualdehyde

PC500542-1G PC500542-250MG PC500542-5G
APOSPC500542-1GEA 7610 CZK
APOSPC500542-1G APOSPC500542-250MG APOSPC500542-5G
2-(Bromomethyl)-4-fluorobenzaldehyde
2-(Bromomethyl)-4-fluorobenzaldehyde
Vzorec: C₈H₆BrFO
MW: 217.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18205513
CAS číslo: 1379346-12-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR