2'-Amino-3'-methoxy-2,2,2-trifluoroacetophenone
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Methoxy-6-(trifluoroacetyl)aniline, 2-Amino-3-(trifluoroacetyl)anisole, 1-(2-Amino-3-methoxyphenyl)-2,2,2-trifluoroethan-1-one
PC50054-1G
PC50054-250MG
APOSPC50054-1GEA
0
CZK
APOSPC50054-1G
APOSPC50054-250MG
2'-Amino-3'-methoxy-2,2,2-trifluoroacetophenone
2'-Amino-3'-methoxy-2,2,2-trifluoroacetophenone
Vzorec:
C₉H₈F₃NO₂ MW: 219,161 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13173364 CAS číslo: 489429-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...