Tisk…

3-(Bromomethyl)-5-fluorobenzonitrile

Dodavatel: Apollo Scientific
Nebezpečí

PC500533-1G PC500533-250MG PC500533-5G
APOSPC500533-1GEA 8270 CZK
APOSPC500533-1G APOSPC500533-250MG APOSPC500533-5G
3-(Bromomethyl)-5-fluorobenzonitrile
3-(Bromomethyl)-5-fluorobenzonitrile
Vzorec: C₈H₅BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD18394044
CAS číslo: 853368-35-1
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR