Tisk…

2-(Bromomethyl)-5-fluorobenzonitrile

Dodavatel: Apollo Scientific
Nebezpečí

PC500527-1G PC500527-25G PC500527-5G
APOSPC500527-1GEA 622 CZK
APOSPC500527-1G APOSPC500527-25G APOSPC500527-5G
2-(Bromomethyl)-5-fluorobenzonitrile
2-(Bromomethyl)-5-fluorobenzonitrile
Vzorec: C₈H₅BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD09261046
CAS číslo: 217661-27-3
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR