Tisk…

3-Bromo-6-chloro-2-fluorobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-Bromo-3-(bromomethyl)-4-chloro-2-fluorobenzene, 6-Chloro-α,3-dibromo-2-fluorotoluene

PC500487-1G PC500487-250MG PC500487-5G
APOSPC500487-1GEA 7440 CZK
APOSPC500487-1G APOSPC500487-250MG APOSPC500487-5G
3-Bromo-6-chloro-2-fluorobenzyl bromide 95%
3-Bromo-6-chloro-2-fluorobenzyl bromide
Vzorec: C₇H₄Br₂ClF
MW: 302,366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27931488
CAS číslo: 886615-32-3
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR