Tisk…

3-Bromo-6-chloro-2-fluorophenacyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-1-(3-bromo-6-chloro-2-fluorophenyl)ethanone

PC500485-1G PC500485-250MG
APOSPC500485-1GEA 25790 CZK
APOSPC500485-1G APOSPC500485-250MG
3-Bromo-6-chloro-2-fluorophenacyl bromide
3-Bromo-6-chloro-2-fluorophenacyl bromide
Vzorec: C₈H₄Br₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD28040504
CAS číslo: 1807121-46-5
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR