Perfluoro(isopropylcyclohexane), tech
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(1,1,1,2,3,3,3-Heptafluoroprop-2-yl)undecafluorocyclohexane, Octadecafluoro(isopropylcyclohexane)
PC50048-25G
PC50048-5G
APOSPC50048-25GEA
21790
CZK
APOSPC50048-25G
APOSPC50048-5G
Perfluoro(isopropylcyclohexane), tech
Perfluoro(isopropylcyclohexane)
Vzorec:
C₉F₁₈ Bod varu: 124…125 °C (749 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00153236 CAS číslo: 423-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...