4-(Bromomethyl)-3-(trifluoromethyl)benzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
PC500478-1G
PC500478-250MG
APOSPC500478-1GEA
11380
CZK
APOSPC500478-1G
APOSPC500478-250MG
4-(Bromomethyl)-3-(trifluoromethyl)benzonitrile
4-(Bromomethyl)-3-(trifluoromethyl)benzonitrile
Vzorec:
C₉H₅BrF₃N MW: 264.042 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22492235 CAS číslo: 853368-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...