4-(Bromomethyl)-3-(trifluoromethyl)benzaldehyde
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC500471-1G
PC500471-5G
PC500471-250MG
APOSPC500471-1GEA
6220
CZK
APOSPC500471-1G
APOSPC500471-5G
APOSPC500471-250MG
4-(Bromomethyl)-3-(trifluoromethyl)benzaldehyde
4-(Bromomethyl)-3-(trifluoromethyl)benzaldehyde
Vzorec:
C₉H₆BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD28014065 CAS číslo: 1416980-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...