Tisk…

5-Bromo-2-fluoro-4-methoxybenzyl alcohol 95%

Dodavatel: Apollo Scientific
Varování

PC500443-1G PC500443-250MG
APOSPC500443-1GEA 17590 CZK
APOSPC500443-1G APOSPC500443-250MG
5-Bromo-2-fluoro-4-methoxybenzyl alcohol 95%
5-Bromo-2-fluoro-4-methoxybenzyl alcohol
Vzorec: C₈H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD11847142
CAS číslo: 1260903-35-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR