Methyl 2-chloro-3-fluorobenzoate
Dodavatel: Apollo Scientific
Synonyms:
2-Chloro-3-fluorobenzoic acid methyl ester
PC500399-1G
PC500399-5G
PC500399-25G
APOSPC500399-1GEA
5100
CZK
APOSPC500399-1G
APOSPC500399-5G
APOSPC500399-25G
Methyl 2-chloro-3-fluorobenzoate
Methyl 2-chloro-3-fluorobenzoate
Vzorec:
C₈H₆ClFO₂ MW: 188,583 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10566821 CAS číslo: 647020-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...