Tisk…

3-Fluoro-4-iodobenzotrifluoride

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-1-iodo-4-(trifluoromethyl)benzene

PC500389-1G PC500389-25G PC500389-5G
APOSPC500389-1GEA 3610 CZK
APOSPC500389-1G APOSPC500389-25G APOSPC500389-5G
3-Fluoro-4-iodobenzotrifluoride
3-Fluoro-4-iodobenzotrifluoride
Vzorec: C₇H₃F₄I
MW: 289.997 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07771984
CAS číslo: 132554-73-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR