Tisk…

2-Fluoro-5-iodo-4-methylanisole

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Fluoro-4-iodo-2-methoxy-5-methylbenzene

PC500322-1G PC500322-250MG
APOSPC500322-1GEA 5790 CZK
APOSPC500322-1G APOSPC500322-250MG
2-Fluoro-5-iodo-4-methylanisole
2-Fluoro-5-iodo-4-methylanisole
Vzorec: C₈H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD27975454
CAS číslo: 1823867-60-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR