Tisk…

2-Amino-6-fluoro-4-(trifluoromethyl)benzonitrile 95%

Dodavatel: Apollo Scientific
Varování

PC500320-1G PC500320-250MG
APOSPC500320-1GEA 11180 CZK
APOSPC500320-1G APOSPC500320-250MG
2-Amino-6-fluoro-4-(trifluoromethyl)benzonitrile 95%
2-Amino-6-fluoro-4-(trifluoromethyl)benzonitrile
Vzorec: C₈H₄F₄N₂
MW: 204.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27975492
CAS číslo: 1807176-08-4
UN: 3439
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR