2-Amino-4-fluoro-6-methylphenol
Dodavatel: Apollo Scientific
PC500315-1G
PC500315-250MG
APOSPC500315-1GEA
7770
CZK
APOSPC500315-1G
APOSPC500315-250MG
2-Amino-4-fluoro-6-methylphenol
2-Amino-4-fluoro-6-methylphenol
Vzorec:
C₇H₈FNO MW: 141,143 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19214250 CAS číslo: 133788-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...