Tisk…

2,6-Difluoro-4-(trifluoromethyl)benzeneboronic acid

Dodavatel: Apollo Scientific
Varování

PC500307-1G PC500307-250MG PC500307-5G
APOSPC500307-1GEA 9430 CZK
APOSPC500307-1G APOSPC500307-250MG APOSPC500307-5G
2,6-Difluoro-4-(trifluoromethyl)benzeneboronic acid
2,6-Difluoro-4-(trifluoromethyl)benzeneboronic acid
Vzorec: C₇H₄BF₅O₂
Storage Temperature: Refrigerator
MDL Number: MFCD27975470
CAS číslo: 2250114-85-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR