Tisk…

6-Fluoro-2-hydroxy-3-nitrobenzoic acid

Dodavatel: Apollo Scientific
Nebezpečí

PC500298-1G PC500298-250MG
APOSPC500298-1GEA 11580 CZK
APOSPC500298-1G APOSPC500298-250MG
6-Fluoro-2-hydroxy-3-nitrobenzoic acid
6-Fluoro-2-hydroxy-3-nitrobenzoic acid
Vzorec: C₇H₄FNO₅
Storage Temperature: Ambient
MDL Number: MFCD27975496
CAS číslo: 1820639-38-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR