Tisk…
5-Bromo-2-chloro-4-fluorophenyl trifluoromethanesulphonate 95%
Dodavatel: Apollo Scientific
Nebezpečí
Synonyms: 5-Bromo-2-chloro-4-fluorophenyl triflate
PC500246-1G PC500246-250MG PC500246-5G
APOSPC500246-1GEA 4440 CZK
APOSPC500246-1G APOSPC500246-250MG APOSPC500246-5G
5-Bromo-2-chloro-4-fluorophenyl trifluoromethanesulphonate 95%
5-Bromo-2-chloro-4-fluorophenyl trifluoromethanesulphonate
Vzorec: C₇H₂BrClF₄O₃S
MW: 357,505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27937161
CAS číslo: 1820619-90-6
UN: 3265
ADR: 8,III