2,4-Difluoro-5-(trifluoromethyl)benzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
PC500230-1G
PC500230-250MG
APOSPC500230-1GEA
11580
CZK
APOSPC500230-1G
APOSPC500230-250MG
2,4-Difluoro-5-(trifluoromethyl)benzonitrile
2,4-Difluoro-5-(trifluoromethyl)benzonitrile
Vzorec:
C₈H₂F₅N Storage Temperature: Ambient |
MDL Number:
MFCD01631621 CAS číslo: 261945-18-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...