2,4-Difluoro-5-hydroxybenzotrifluoride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,4-Difluoro-5-(trifluoromethyl)phenol
PC500224-1G
PC500224-250MG
APOSPC500224-1GEA
15710
CZK
APOSPC500224-1G
APOSPC500224-250MG
2,4-Difluoro-5-hydroxybenzotrifluoride
2,4-Difluoro-5-hydroxybenzotrifluoride
Vzorec:
C₇H₃F₅O MW: 198,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27937185 CAS číslo: 1632283-20-5 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...