2,6-Difluoro-3-(trifluoromethyl)benzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC500223-1G
PC500223-250MG
APOSPC500223-1GEA
12010
CZK
APOSPC500223-1G
APOSPC500223-250MG
2,6-Difluoro-3-(trifluoromethyl)benzaldehyde
2,6-Difluoro-3-(trifluoromethyl)benzaldehyde
Vzorec:
C₈H₃F₅O MW: 210.101 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27937180 CAS číslo: 134099-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...