Tisk…

4-Bromo-3-fluoro-2-methoxyaniline

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-3-fluoro-o-anisidine, 6-Amino-3-bromo-2-fluoroanisole

PC500208-1G PC500208-5G PC500208-250MG
APOSPC500208-1GEA 8200 CZK
APOSPC500208-1G APOSPC500208-5G APOSPC500208-250MG
4-Bromo-3-fluoro-2-methoxyaniline
4-Bromo-3-fluoro-2-methoxyaniline
Vzorec: C₇H₇BrFNO
MW: 220,039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22570403
CAS číslo: 1137869-95-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR