Tisk…

3-Chloro-2-(trifluoromethyl)phenol

Dodavatel: Apollo Scientific

PC500171-1G PC500171-250MG PC500171-5G
APOSPC500171-1GEA 6910 CZK
APOSPC500171-1G APOSPC500171-250MG APOSPC500171-5G
3-Chloro-2-(trifluoromethyl)phenol
3-Chloro-2-(trifluoromethyl)phenol
Vzorec: C₇H₄ClF₃O
MW: 196,554 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17000111
CAS číslo: 106877-34-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR