4-Bromo-2-fluorobenzylsulphonyl chloride
Dodavatel: Apollo Scientific
PC50015-1G
PC50015-250MG
APOSPC50015-1GEA
10350
CZK
APOSPC50015-1G
APOSPC50015-250MG
4-Bromo-2-fluorobenzylsulphonyl chloride
4-Bromo-2-fluorobenzylsulphonyl chloride
Vzorec:
C₇H₅BrClFO₂S Storage Temperature: Ambient |
MDL Number:
MFCD11976108 CAS číslo: 1178830-06-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...