4-Chloro-3-(trifluoromethyl)phenyl trifluoromethanesulphonate 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-5-{[(trifluoromethyl)sulphonyl]oxy}benzotrifluoride, 4-Chloro-3-(trifluoromethyl)phenyl triflate
PC500097-1G
PC500097-250MG
APOSPC500097-1GEA
13890
CZK
APOSPC500097-1G
APOSPC500097-250MG
4-Chloro-3-(trifluoromethyl)phenyl trifluoromethanesulphonate 97%
4-Chloro-3-(trifluoromethyl)phenyl trifluoromethanesulphonate
Vzorec:
C₈H₃ClF₆O₃S MW: 328,616 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD25541859 CAS číslo: 1446016-91-6 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...