Tisk…
2-Fluoro-5-(trifluoromethyl)phenyl trifluoromethanesulphonate 95%
Dodavatel: Apollo Scientific
Nebezpečí
Synonyms: 2-Fluoro-5-(trifluoromethyl)phenyl triflate, 4-Fluoro-3-{[(trifluoromethyl)sulphonyl]oxy}benzotrifluoride
PC500070-1G PC500070-250MG
APOSPC500070-1GEA 5790 CZK
APOSPC500070-1G APOSPC500070-250MG
2-Fluoro-5-(trifluoromethyl)phenyl trifluoromethanesulphonate 95%
2-Fluoro-5-(trifluoromethyl)phenyl trifluoromethanesulphonate
Vzorec: C₈H₃F₇O₃S
MW: 312.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25541832
CAS číslo: 1446016-79-0
UN: 3265
ADR: 8 III