Tisk…

Methyl 2H,2H,3H,3H-perfluoroheptanoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 4,4,5,5,6,6,7,7,7-nonafluoroheptanoate

PC50003-1G PC50003-250MG
APOSPC50003-1GEA 21790 CZK
APOSPC50003-1G APOSPC50003-250MG
Methyl 2H,2H,3H,3H-perfluoroheptanoate
Methyl 2H,2H,3H,3H-perfluoroheptanoate
Vzorec: C₈H₇F₉O₂
MW: 306,126 g/mol
Bod varu: 53…54 °C (9 mm Hg)
Hustota: 1,1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD17676091
CAS číslo: 132424-36-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR