4-(Trifluoromethyl)isatoic anhydride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
7-(Trifluoromethyl)-2H-3,1-benzoxazine-2,4(1H)-dione, 1,4-Dihydro-2,4-dioxo-7-(trifluoromethyl)-2H-3,1-benzoxazine, 7-(Trifluoromethyl)-1H-benzo[d][1,3]oxazine-2,4-dione
PC500015-1G
PC500015-250MG
PC500015-5G
APOSPC500015-1GEA
2150
CZK
APOSPC500015-1G
APOSPC500015-250MG
APOSPC500015-5G
4-(Trifluoromethyl)isatoic anhydride
4-(Trifluoromethyl)isatoic anhydride
Vzorec:
C₉H₄F₃NO₃ MW: 231,128 g/mol Bod tání: 240…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD21946140 CAS číslo: 97928-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...