Tisk…

2,3-Difluoro-4-(trifluoromethyl)benzonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Cyano-2,3-difluorobenzotrifluoride

PC4993-1G PC4993-250MG PC4993-5G
APOSPC4993-1GEA 7340 CZK
APOSPC4993-1G APOSPC4993-250MG APOSPC4993-5G
2,3-Difluoro-4-(trifluoromethyl)benzonitrile 98%
2,3-Difluoro-4-(trifluoromethyl)benzonitrile
Vzorec: C₈H₂F₅N
MW: 207.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236258
CAS číslo: 240122-24-1
UN: 3276
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR