Tisk…

4-(2,2,2-Trifluoroethylamino)benzoic acid 95%

Dodavatel: Apollo Scientific

PC499030-1G PC499030-250MG
APOSPC499030-1GEA 16340 CZK
APOSPC499030-1G APOSPC499030-250MG
4-(2,2,2-Trifluoroethylamino)benzoic acid 95%
4-(2,2,2-Trifluoroethylamino)benzoic acid
Vzorec: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD16692383
CAS číslo: 892878-66-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR