Tisk…

3-(2,2,2-Trifluoroethylamino)benzoic acid 95%

Dodavatel: Apollo Scientific

PC499029-1G PC499029-250MG
APOSPC499029-1GEA 17360 CZK
APOSPC499029-1G APOSPC499029-250MG
3-(2,2,2-Trifluoroethylamino)benzoic acid 95%
3-(2,2,2-Trifluoroethylamino)benzoic acid
Vzorec: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD20525970
CAS číslo: 1215366-23-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR