7-Bromo-2-(trifluoromethyl)indole 95%
Dodavatel: Apollo Scientific
PC499020-1G
PC499020-250MG
PC499020-5G
APOSPC499020-1GEA
17300
CZK
APOSPC499020-1G
APOSPC499020-250MG
APOSPC499020-5G
7-Bromo-2-(trifluoromethyl)indole 95%
7-Bromo-2-(trifluoromethyl)indole
Vzorec:
C₉H₅BrF₃N MW: 264,042 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30721261 CAS číslo: 1779887-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...