5-Bromo-2-(trifluoromethyl)indole
Dodavatel: Apollo Scientific
PC499019-10G
PC499019-1G
PC499019-5G
APOSPC499019-10GEA
74390
CZK
APOSPC499019-10G
APOSPC499019-1G
APOSPC499019-5G
5-Bromo-2-(trifluoromethyl)indole
5-Bromo-2-(trifluoromethyl)indole
Vzorec:
C₉H₅BrF₃N MW: 264,042 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20230421 CAS číslo: 837392-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...