3,5-Difluoro-2-nitrophenylacetonitrile
Dodavatel: Apollo Scientific
PC499018-1G
PC499018-250MG
APOSPC499018-1GEA
11180
CZK
APOSPC499018-1G
APOSPC499018-250MG
3,5-Difluoro-2-nitrophenylacetonitrile
3,5-Difluoro-2-nitrophenylacetonitrile
Vzorec:
C₈H₄F₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD24221999 CAS číslo: 1550550-56-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...